Assay Details
| Gene Symbol: sgk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047834.4 | NP_001041299.1 | 9 - 10 | 991 | 95 | |
| NM_001381177.1 | NP_001367134.1 | 9 - 10 | 1125 | 95 | ||
More Information
Set Membership: |
|

sgk-1
Genomic Map