Assay Details
| Gene Symbol: taf-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270343.3 | NP_001257272.1 | 2 - 3 | 305 | 77 | |
| NM_001347279.5 | NP_001334235.1 | 1 - 2 | 206 | 77 | ||
More Information
Set Membership: |
|

taf-3
Genomic Map