Assay Details
| Gene Symbol: ace-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078259.8 | NP_510660.1 | 4 - 5 | 793 | 69 | |
| GenBank mRNA | X75331.1 | - | 857 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ace-1| Gene Symbol: ace-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078259.8 | NP_510660.1 | 4 - 5 | 793 | 69 | |
| GenBank mRNA | X75331.1 | - | 857 | 69 | ||
Set Membership: |
|