Assay Details
| Gene Symbol: ser-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029557.5 | NP_001024728.1 | 1 - 2 | 63 | 55 | |
| GenBank mRNA | AF031414.1 | - | 1 - 2 | 107 | 55 | |
More Information
Set Membership: |
|

ser-1
Genomic Map