Assay Details
| Gene Symbol: ser-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029557.5 | NP_001024728.1 | 3 - 4 | 1077 | 60 | |
| NM_001029558.5 | NP_001024729.1 | 2 - 3 | 933 | 60 | ||
| GenBank mRNA | AF031414.1 | - | 3 - 4 | 1121 | 60 | |
| AF031415.1 | - | 2 - 3 | 1035 | 60 | ||
More Information
Set Membership: |
|

ser-1
Genomic Map