Assay Details
| Gene Symbol: ser-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029557.5 | NP_001024728.1 | 5 - 6 | 1580 | 86 | |
| NM_001029558.5 | NP_001024729.1 | 4 - 5 | 1436 | 86 | ||
| GenBank mRNA | AF031414.1 | - | 5 - 6 | 1624 | 86 | |
| AF031415.1 | - | 4 - 5 | 1538 | 86 | ||
More Information
Set Membership: |
|

ser-1
Genomic Map