Assay Details
| Gene Symbol: ain-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078286.8 | NP_510687.2 | 6 - 7 | 1875 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ain-1| Gene Symbol: ain-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078286.8 | NP_510687.2 | 6 - 7 | 1875 | 102 | |
Set Membership: |
|