Assay Details
| Gene Symbol: csk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306233.3 | NP_001293162.1 | 2 - 3 | 174 | 81 | |
| NM_001380379.1 | NP_001367885.1 | 1 - 2 | 89 | 81 | ||
| GenBank mRNA | AB096875.1 | - | 1 - 2 | 76 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001380379.1 transcripts.
Set Membership: |
|

csk-1
Genomic Map