Assay Details
| Gene Symbol: wwp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170794.7 | NP_740775.1 | 4 - 5 | 825 | 64 | |
| NM_171831.4 | NP_740776.1 | 4 - 5 | 825 | 64 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

wwp-1| Gene Symbol: wwp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170794.7 | NP_740775.1 | 4 - 5 | 825 | 64 | |
| NM_171831.4 | NP_740776.1 | 4 - 5 | 825 | 64 | ||
Set Membership: |
|