Assay Details
| Gene Symbol: wwp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306342.3 | NP_001293271.1 | 3 - 4 | 759 | 103 | |
| NM_001306343.3 | NP_001293272.1 | 3 - 4 | 753 | 103 | ||
| NM_170794.7 | NP_740775.1 | 7 - 8 | 1636 | 103 | ||
| NM_171831.4 | NP_740776.1 | 7 - 8 | 1630 | 103 | ||
More Information
Set Membership: |
|

wwp-1
Genomic Map