Assay Details
| Gene Symbol: rpl-17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170799.6 | NP_740781.1 | 2 - 3 | 450 | 83 | |
| Gene Symbol: Y48G8AL.7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381094.2 | NP_001368003.1 | 2 - 3 | 425 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001381094.2 transcripts.
Set Membership: |
|

rpl-17
Genomic Map