Assay Details
| Gene Symbol: C53H9.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170804.6 | NP_740787.2 | 3 - 4 | 740 | 88 | |
| NR_131348.1 | 2 - 3 | 782 | 88 | |||
| NR_131349.1 | 3 - 4 | 715 | 88 | |||
| NR_131350.1 | 3 - 4 | 715 | 88 | |||
| NR_151555.1 | 3 - 4 | 740 | 88 | |||
| Gene Symbol: rpl-27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381283.2 | NP_001367687.1 | 2 - 3 | 789 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001381283.2 transcripts.
Set Membership: |
|

C53H9.2
Genomic Map