Assay Details
| Gene Symbol: spe-48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306469.2 | NP_001293398.1 | 6 - 7 | 871 | 117 | |
| NM_001306470.3 | NP_001293399.1 | 5 - 6 | 752 | 117 | ||
| NM_001306471.3 | NP_001293400.1 | 5 - 6 | 827 | 117 | ||
| NM_001306472.3 | NP_001293401.1 | 1 - 2 | 86 | 117 | ||
| NM_170811.6 | NP_740794.1 | 6 - 7 | 946 | 117 | ||
More Information
Set Membership: |
|

spe-48
Genomic Map