Assay Details
| Gene Symbol: ubr-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318271.2 | NP_001305200.1 | 16 - 17 | 9518 | 128 | |
| NM_001382970.1 | 16 - 17 | 9547 | 128 | |||
| NM_059006.3 | NP_491407.2 | 14 - 15 | 9497 | 128 | ||
| NR_133620.1 | 13 - 14 | 10254 | 128 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001382970.1 transcripts.
Set Membership: |
|

ubr-4
Genomic Map