Assay Details
| Gene Symbol: fhod-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313484.2 | NP_001300413.1 | 14 - 15 | 3394 | 90 | |
| NM_001313485.3 | NP_001300414.1 | 16 - 17 | 3918 | 90 | ||
| NM_001313486.4 | NP_001300415.1 | 15 - 16 | 3447 | 90 | ||
| NM_001313487.3 | NP_001300416.1 | 15 - 16 | 4923 | 90 | ||
| NM_001313488.4 | NP_001300417.1 | 14 - 15 | 4876 | 90 | ||
| NM_001313489.3 | NP_001300418.1 | 14 - 15 | 4449 | 90 | ||
| NM_001313490.3 | NP_001300419.1 | 13 - 14 | 4398 | 90 | ||
| NM_059112.4 | NP_491513.2 | 15 - 16 | 3868 | 90 | ||
| GenBank mRNA | AB084086.1 | - | 15 - 16 | 3890 | 90 | |
More Information
Set Membership: |
|

fhod-1
Genomic Map