Assay Details
| Gene Symbol: fhod-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313484.2 | NP_001300413.1 | 1 - 2 | 137 | 144 | |
| NM_001313485.3 | NP_001300414.1 | 1 - 2 | 136 | 144 | ||
| NM_001313486.4 | NP_001300415.1 | 1 - 2 | 139 | 144 | ||
| NM_059112.4 | NP_491513.2 | 1 - 2 | 137 | 144 | ||
| GenBank mRNA | AB084086.1 | - | 1 - 2 | 159 | 144 | |
More Information
Set Membership: |
|

fhod-1
Genomic Map