Assay Details
| Gene Symbol: fhod-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313484.2 | NP_001300413.1 | 5 - 6 | 638 | 117 | |
| NM_001313488.4 | NP_001300417.1 | 4 - 5 | 1646 | 117 | ||
| NM_001313490.3 | NP_001300419.1 | 4 - 5 | 1642 | 117 | ||
| NM_059112.4 | NP_491513.2 | 5 - 6 | 638 | 117 | ||
| GenBank mRNA | AB084086.1 | - | 5 - 6 | 660 | 117 | |
More Information
Set Membership: |
|

fhod-1
Genomic Map