Assay Details
| Gene Symbol: tom-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313498.3 | NP_001300427.1 | 13 - 14 | 2116 | 103 | |
| NM_001313499.3 | NP_001300428.1 | 13 - 14 | 2116 | 103 | ||
| NM_001313500.3 | NP_001300429.1 | 13 | 2009 | 103 | ||
More Information
Set Membership: |
|

tom-1
Genomic Map