Assay Details
| Gene Symbol: tom-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313498.3 | NP_001300427.1 | 14 - 15 | 2605 | 73 | |
| NM_001313499.3 | NP_001300428.1 | 14 - 15 | 2605 | 73 | ||
| NM_001313500.3 | NP_001300429.1 | 14 | 2498 | 73 | ||
More Information
Set Membership: |
|

tom-1
Genomic Map