Assay Details
| Gene Symbol: tom-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313499.3 | NP_001300428.1 | 16 - 17 | 2870 | 113 | |
| NM_059222.7 | NP_491623.3 | 16 - 17 | 2504 | 113 | ||
| GenBank mRNA | AY912102.1 | - | 16 - 17 | 2397 | 113 | |
More Information
Set Membership: |
|

tom-1
Genomic Map