Assay Details
| Gene Symbol: tom-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129022.5 | NP_001122494.1 | 18 - 19 | 2660 | 59 | |
| NM_001129023.2 | NP_001122495.1 | 18 - 19 | 2678 | 59 | ||
| NM_001313498.3 | NP_001300427.1 | 18 - 19 | 3027 | 59 | ||
| NM_001313499.3 | NP_001300428.1 | 17 - 18 | 3015 | 59 | ||
| NM_001313500.3 | NP_001300429.1 | 18 | 2935 | 59 | ||
| NM_059222.7 | NP_491623.3 | 17 - 18 | 2649 | 59 | ||
| GenBank mRNA | AY912102.1 | - | 17 - 18 | 2542 | 59 | |
| AY997305.1 | - | 18 - 19 | 2554 | 59 | ||
| AY997306.1 | - | 18 - 19 | 2569 | 59 | ||
More Information
Set Membership: |
|

tom-1
Genomic Map