Assay Details
| Gene Symbol: tom-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129022.5 | NP_001122494.1 | 19 - 20 | 2968 | 126 | |
| NM_001129023.2 | NP_001122495.1 | 19 - 20 | 2986 | 126 | ||
| NM_001313498.3 | NP_001300427.1 | 19 - 20 | 3335 | 126 | ||
| NM_001313499.3 | NP_001300428.1 | 18 - 19 | 3323 | 126 | ||
| NM_001313500.3 | NP_001300429.1 | 19 | 3243 | 126 | ||
| NM_059222.7 | NP_491623.3 | 18 - 19 | 2957 | 126 | ||
| GenBank mRNA | AY912102.1 | - | 18 - 19 | 2850 | 126 | |
| AY997305.1 | - | 19 - 20 | 2862 | 126 | ||
| AY997306.1 | - | 19 - 20 | 2877 | 126 | ||
More Information
Set Membership: |
|

tom-1
Genomic Map