Assay Details
| Gene Symbol: tom-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129022.5 | NP_001122494.1 | 4 - 5 | 559 | 147 | |
| NM_001129023.2 | NP_001122495.1 | 4 - 5 | 562 | 147 | ||
| NM_001313498.3 | NP_001300427.1 | 4 - 5 | 560 | 147 | ||
| NM_001313499.3 | NP_001300428.1 | 4 - 5 | 560 | 147 | ||
| NM_001313500.3 | NP_001300429.1 | 4 | 453 | 147 | ||
| NM_059222.7 | NP_491623.3 | 4 - 5 | 560 | 147 | ||
| GenBank mRNA | AY912102.1 | - | 4 - 5 | 453 | 147 | |
| AY997305.1 | - | 4 - 5 | 453 | 147 | ||
| AY997306.1 | - | 4 - 5 | 453 | 147 | ||
More Information
Set Membership: |
|

tom-1
Genomic Map