Assay Details
| Gene Symbol: tns-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318270.3 | NP_001305199.1 | 12 - 13 | 2218 | 104 | |
| NM_001383115.1 | 13 - 14 | 2302 | 104 | |||
| NM_059236.5 | NP_491637.2 | 14 - 15 | 2341 | 104 | ||
| NM_170867.5 | NP_740854.1 | 4 - 5 | 1238 | 104 | ||
More Information
Set Membership: |
|

tns-1
Genomic Map