Assay Details
| Gene Symbol: tns-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318270.3 | NP_001305199.1 | 13 - 14 | 2412 | 90 | |
| NM_001318273.3 | NP_001305202.1 | 1 - 2 | 171 | 90 | ||
| NM_001383115.1 | 14 - 15 | 2496 | 90 | |||
| NM_059236.5 | NP_491637.2 | 15 - 16 | 2535 | 90 | ||
| NM_170867.5 | NP_740854.1 | 5 - 6 | 1432 | 90 | ||
More Information
Set Membership: |
|

tns-1
Genomic Map