Assay Details
| Gene Symbol: tns-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318270.3 | NP_001305199.1 | 16 - 17 | 3016 | 79 | |
| NM_001318273.3 | NP_001305202.1 | 4 - 5 | 775 | 79 | ||
| NM_001383115.1 | 17 - 18 | 3100 | 79 | |||
| NM_059236.5 | NP_491637.2 | 18 - 19 | 3139 | 79 | ||
| NM_170867.5 | NP_740854.1 | 8 - 9 | 2036 | 79 | ||
More Information
Set Membership: |
|

tns-1
Genomic Map