Assay Details
| Gene Symbol: nsf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083134.4 | NP_001076603.1 | 10 - 11 | 2082 | 118 | |
| NM_001083135.5 | NP_001076604.1 | 8 - 9 | 1861 | 118 | ||
More Information
Set Membership: |
|

nsf-1
Genomic Map