Assay Details
| Gene Symbol: nsf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083134.4 | NP_001076603.1 | 3 - 4 | 314 | 93 | |
| NM_001083135.5 | NP_001076604.1 | 1 - 2 | 93 | 93 | ||
More Information
Set Membership: |
|

nsf-1
Genomic Map