Assay Details
| Gene Symbol: hmr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026479.4 | NP_001021650.1 | 21 - 22 | 5293 | 61 | |
| GenBank mRNA | AJ307058.1 | - | 21 - 22 | 5293 | 61 | |
More Information
Set Membership: |
|

hmr-1
Genomic Map