Assay Details
| Gene Symbol: hmr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026479.4 | NP_001021650.1 | 26 - 27 | 6536 | 88 | |
| NM_001264632.3 | NP_001251561.1 | 3 - 4 | 860 | 88 | ||
| NM_001381215.1 | NP_001366962.1 | 6 - 7 | 1701 | 88 | ||
| NM_001381216.1 | NP_001368105.1 | 1 - 2 | 470 | 88 | ||
| GenBank mRNA | AJ307058.1 | - | 26 - 27 | 6536 | 88 | |
More Information
Set Membership: |
|

hmr-1
Genomic Map