Assay Details
| Gene Symbol: slc-30A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170935.8 | NP_740931.2 | 5 - 6 | 1190 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

slc-30A5| Gene Symbol: slc-30A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170935.8 | NP_740931.2 | 5 - 6 | 1190 | 66 | |
Set Membership: |
|