Assay Details
| Gene Symbol: kin-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381269.1 | NP_001368699.1 | 9 - 10 | 1583 | 73 | |
| NM_001381270.1 | NP_001367024.1 | 6 - 7 | 1023 | 73 | ||
| NM_001381271.1 | NP_001367023.1 | 6 - 7 | 909 | 73 | ||
| NM_001381272.1 | NP_001366905.1 | 6 - 7 | 1057 | 73 | ||
| NM_001381273.1 | NP_001367025.1 | 6 - 7 | 990 | 73 | ||
| NM_001381274.1 | NP_001367022.1 | 6 - 7 | 934 | 73 | ||
| NM_170965.3 | NP_740961.1 | 6 - 7 | 1143 | 73 | ||
More Information
Set Membership: |
|

kin-1
Genomic Map