Assay Details
| Gene Symbol: shc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383823.1 | 7 - 8 | 1054 | 123 | ||
| NM_001383824.1 | 7 - 8 | 1040 | 123 | |||
| NM_062514.4 | NP_494915.2 | 7 - 8 | 1226 | 123 | ||
More Information
Set Membership: |
|

shc-2
Genomic Map