Assay Details
| Gene Symbol: crh-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306557.3 | NP_001293486.1 | 3 - 4 | 337 | 84 | |
| NM_170987.8 | NP_740985.2 | 2 - 3 | 369 | 84 | ||
| NM_170988.7 | NP_740986.2 | 6 - 7 | 683 | 84 | ||
| NM_170989.11 | NP_740987.4 | 5 - 6 | 923 | 84 | ||
More Information
Set Membership: |
|

crh-2
Genomic Map