Assay Details
| Gene Symbol: crh-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306557.3 | NP_001293486.1 | 1 - 2 | 123 | 77 | |
| NM_170988.7 | NP_740986.2 | 4 - 5 | 469 | 77 | ||
| NM_170989.11 | NP_740987.4 | 3 - 4 | 709 | 77 | ||
More Information
Set Membership: |
|

crh-2
Genomic Map