Assay Details
| Gene Symbol: unc-104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026870.7 | NP_001022041.2 | 10 - 11 | 2087 | 172 | |
| NM_171017.9 | NP_741019.3 | 10 - 11 | 2087 | 172 | ||
More Information
Set Membership: |
|

unc-104
Genomic Map