Assay Details
| Gene Symbol: cam-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026736.5 | NP_001021907.1 | 2 - 3 | 211 | 90 | |
| NM_001026737.5 | NP_001021908.1 | 2 - 3 | 243 | 90 | ||
| NM_001383915.1 | 2 - 3 | 202 | 90 | |||
| GenBank mRNA | AJ132946.1 | - | 2 - 3 | 210 | 90 | |
| AJ132947.1 | - | 1 - 2 | 131 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383915.1 transcripts.
Set Membership: |
|

cam-1
Genomic Map