Assay Details
| Gene Symbol: cam-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026736.5 | NP_001021907.1 | 5 - 6 | 881 | 66 | |
| NM_001026737.5 | NP_001021908.1 | 5 - 6 | 913 | 66 | ||
| NM_001383915.1 | 5 - 6 | 876 | 66 | |||
| GenBank mRNA | AJ132946.1 | - | 5 - 6 | 880 | 66 | |
| AJ132947.1 | - | 4 - 5 | 801 | 66 | ||
More Information
Set Membership: |
|

cam-1
Genomic Map