Assay Details
| Gene Symbol: cam-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026736.5 | NP_001021907.1 | 6 - 7 | 1023 | 108 | |
| NM_001026737.5 | NP_001021908.1 | 6 - 7 | 1055 | 108 | ||
| NM_001383915.1 | 6 - 7 | 1018 | 108 | |||
| GenBank mRNA | AJ132946.1 | - | 6 - 7 | 1022 | 108 | |
| AJ132947.1 | - | 5 - 6 | 943 | 108 | ||
More Information
Set Membership: |
|

cam-1
Genomic Map