Assay Details
| Gene Symbol: kin-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171029.8 | NP_741034.2 | 1 - 2 | 168 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

kin-15| Gene Symbol: kin-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171029.8 | NP_741034.2 | 1 - 2 | 168 | 112 | |
Set Membership: |
|