Assay Details
| Gene Symbol: taf-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027342.4 | NP_001022513.1 | 2 - 3 | 318 | 84 | |
| NM_001027343.5 | NP_001022514.1 | 2 - 3 | 324 | 84 | ||
More Information
Set Membership: |
|

taf-8
Genomic Map