Assay Details
| Gene Symbol: taf-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027342.4 | NP_001022513.1 | 3 - 4 | 481 | 147 | |
| NM_001027343.5 | NP_001022514.1 | 3 - 4 | 487 | 147 | ||
More Information
Set Membership: |
|

taf-8
Genomic Map