Assay Details
| Gene Symbol: ptp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026771.3 | NP_001021942.1 | 10 - 11 | 1606 | 78 | |
| NM_001267341.4 | NP_001254270.1 | 7 - 8 | 1061 | 78 | ||
| NM_001276741.3 | NP_001263670.1 | 11 - 12 | 1678 | 78 | ||
| NM_001276742.3 | NP_001263671.1 | 11 - 12 | 1810 | 78 | ||
| NM_001276743.3 | NP_001263672.1 | 10 - 11 | 1738 | 78 | ||
| NM_001377858.3 | NP_001364824.1 | 11 - 12 | 1678 | 78 | ||
| NM_001377859.2 | NP_001364823.1 | 10 - 11 | 1606 | 78 | ||
| NM_001377860.2 | NP_001364658.1 | 8 - 9 | 1093 | 78 | ||
| NM_001377861.2 | NP_001364506.1 | 8 - 9 | 1131 | 78 | ||
| NM_001377862.2 | NP_001364507.1 | 7 - 8 | 1021 | 78 | ||
More Information
Set Membership: |
|

ptp-3
Genomic Map