Assay Details
| Gene Symbol: ptp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026771.3 | NP_001021942.1 | 12 - 13 | 1878 | 81 | |
| NM_001267341.4 | NP_001254270.1 | 9 - 10 | 1333 | 81 | ||
| NM_001276741.3 | NP_001263670.1 | 13 - 14 | 1950 | 81 | ||
| NM_001276742.3 | NP_001263671.1 | 13 - 14 | 2082 | 81 | ||
| NM_001276743.3 | NP_001263672.1 | 12 - 13 | 2010 | 81 | ||
| NM_001377860.2 | NP_001364658.1 | 10 - 11 | 1365 | 81 | ||
More Information
Set Membership: |
|

ptp-3
Genomic Map