Assay Details
| Gene Symbol: ptp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026771.3 | NP_001021942.1 | 13 - 14 | 2248 | 69 | |
| NM_001267341.4 | NP_001254270.1 | 10 - 11 | 1703 | 69 | ||
| NM_001276741.3 | NP_001263670.1 | 14 - 15 | 2320 | 69 | ||
| NM_001276742.3 | NP_001263671.1 | 14 - 15 | 2452 | 69 | ||
| NM_001276743.3 | NP_001263672.1 | 13 - 14 | 2380 | 69 | ||
| NM_001377860.2 | NP_001364658.1 | 11 - 12 | 1735 | 69 | ||
More Information
Set Membership: |
|

ptp-3
Genomic Map