Assay Details
| Gene Symbol: ptp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026771.3 | NP_001021942.1 | 14 - 15 | 2297 | 69 | |
| NM_001026772.5 | NP_001021943.1 | 2 - 3 | 250 | 69 | ||
| NM_001267341.4 | NP_001254270.1 | 11 - 12 | 1752 | 69 | ||
| NM_001276741.3 | NP_001263670.1 | 15 - 16 | 2369 | 69 | ||
| NM_001276742.3 | NP_001263671.1 | 15 - 16 | 2501 | 69 | ||
| NM_001276743.3 | NP_001263672.1 | 14 - 15 | 2429 | 69 | ||
| NM_001377858.3 | NP_001364824.1 | 15 - 16 | 2039 | 69 | ||
| NM_001377859.2 | NP_001364823.1 | 14 - 15 | 1967 | 69 | ||
| NM_001377860.2 | NP_001364658.1 | 12 - 13 | 1784 | 69 | ||
| NM_001377861.2 | NP_001364506.1 | 12 - 13 | 1492 | 69 | ||
| NM_001377862.2 | NP_001364507.1 | 11 - 12 | 1382 | 69 | ||
More Information
Set Membership: |
|

ptp-3
Genomic Map