Assay Details
| Gene Symbol: ptp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026771.3 | NP_001021942.1 | 16 - 17 | 2776 | 71 | |
| NM_001026772.5 | NP_001021943.1 | 4 - 5 | 729 | 71 | ||
| NM_001267341.4 | NP_001254270.1 | 13 - 14 | 2231 | 71 | ||
| NM_001276741.3 | NP_001263670.1 | 17 - 18 | 2848 | 71 | ||
| NM_001276742.3 | NP_001263671.1 | 17 - 18 | 2980 | 71 | ||
| NM_001276743.3 | NP_001263672.1 | 16 - 17 | 2908 | 71 | ||
| NM_001377858.3 | NP_001364824.1 | 17 - 18 | 2518 | 71 | ||
| NM_001377859.2 | NP_001364823.1 | 16 - 17 | 2446 | 71 | ||
| NM_001377860.2 | NP_001364658.1 | 14 - 15 | 2263 | 71 | ||
| NM_001377861.2 | NP_001364506.1 | 14 - 15 | 1971 | 71 | ||
| NM_001377862.2 | NP_001364507.1 | 13 - 14 | 1861 | 71 | ||
More Information
Set Membership: |
|

ptp-3
Genomic Map