Assay Details
| Gene Symbol: ptp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026771.3 | NP_001021942.1 | 18 - 19 | 3030 | 62 | |
| NM_001026772.5 | NP_001021943.1 | 6 - 7 | 983 | 62 | ||
| NM_001267341.4 | NP_001254270.1 | 15 - 16 | 2485 | 62 | ||
| NM_001276741.3 | NP_001263670.1 | 19 - 20 | 3102 | 62 | ||
| NM_001276742.3 | NP_001263671.1 | 19 - 20 | 3234 | 62 | ||
| NM_001276743.3 | NP_001263672.1 | 18 - 19 | 3162 | 62 | ||
| NM_001377858.3 | NP_001364824.1 | 19 - 20 | 2772 | 62 | ||
| NM_001377859.2 | NP_001364823.1 | 18 - 19 | 2700 | 62 | ||
| NM_001377860.2 | NP_001364658.1 | 16 - 17 | 2517 | 62 | ||
| NM_001377861.2 | NP_001364506.1 | 16 - 17 | 2225 | 62 | ||
| NM_001377862.2 | NP_001364507.1 | 15 - 16 | 2115 | 62 | ||
More Information
Set Membership: |
|

ptp-3
Genomic Map