Assay Details
| Gene Symbol: ptp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026771.3 | NP_001021942.1 | 1 - 2 | 133 | 99 | |
| NM_001276741.3 | NP_001263670.1 | 1 - 2 | 133 | 99 | ||
| NM_001377858.3 | NP_001364824.1 | 1 - 2 | 133 | 99 | ||
| NM_001377859.2 | NP_001364823.1 | 1 - 2 | 133 | 99 | ||
More Information
Set Membership: |
|

ptp-3
Genomic Map