Assay Details
| Gene Symbol: ptp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026771.3 | NP_001021942.1 | 20 - 21 | 3239 | 66 | |
| NM_001026772.5 | NP_001021943.1 | 8 - 9 | 1192 | 66 | ||
| NM_001026773.5 | NP_001021944.1 | 2 - 3 | 350 | 66 | ||
| NM_001267341.4 | NP_001254270.1 | 17 - 18 | 2694 | 66 | ||
| NM_001276741.3 | NP_001263670.1 | 21 - 22 | 3311 | 66 | ||
| NM_001276742.3 | NP_001263671.1 | 21 - 22 | 3443 | 66 | ||
| NM_001276743.3 | NP_001263672.1 | 20 - 21 | 3371 | 66 | ||
| NM_001276744.4 | NP_001263673.1 | 2 - 3 | 998 | 66 | ||
| NM_001377858.3 | NP_001364824.1 | 21 - 22 | 2981 | 66 | ||
| NM_001377859.2 | NP_001364823.1 | 20 - 21 | 2909 | 66 | ||
| NM_001377860.2 | NP_001364658.1 | 18 - 19 | 2726 | 66 | ||
| NM_001377861.2 | NP_001364506.1 | 18 - 19 | 2434 | 66 | ||
| NM_001377862.2 | NP_001364507.1 | 17 - 18 | 2324 | 66 | ||
More Information
Set Membership: |
|

ptp-3
Genomic Map